Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLUT1 Rabbit anti-Human, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA137782

Catalog No. 01-673-521


Add to cart

Description

Description

This antibody is predicted to react with bovine, chicken, mouse, porcine, rabbit and rat based on sequence homology. Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is esophageal or breast carcinoma.

Glucose is fundamental to the metabolism of mammalian cells. Several glucose transporter protein (Glut) isoforms have been identified and shown to function in response to insulin and IGF-1 induced signaling. GLUT-1 is detectable in many human tissues including those of the colon, lung, stomach, esophagus, and breast. GLUT-1 immunoreactivity in some cancers, including trans carcinoma of the urinary bladder, has been associated with aggressive behavior.
Specifications

Specifications

GLUT1
Polyclonal
PBS with 1% BSA and 0.1% sodium azide; pH 7.4
P11166, P11167
DYT17, DYT18, GLUT, GLUT1, GLUT1DS, PED, GLUT-1, glucose transporter type 1, erythrocyte/brain, hepG2 glucose transporter, solute carrier family 2, facilitated glucose transporter member 1
Rabbit
IgG
1 mL
-20° C, Avoid Freeze/Thaw Cycles
Primary
24778, 6513
Immunohistochemistry (Paraffin)
Unconjugated
SLC2A1
Liquid
SLC2A1
Synthetic peptide derived from C-terminus of human GLUT-1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit