Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLUT10 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA14372

Catalog No. PA14372


Add to cart

Description

Description

PA1-4372 detects GLUT10 from human samples. PA1-4372 has been successfully used in Western blot procedures. PA1-4372 immunizing peptide corresponds to amino acid residues 367-385 from human GLUT10 protein.

Non-insulin-dependent diabetes mellitus (NIDDM) is a multifactoral disease with both environmental and genetics causes. Genome-wide screening procedures have identified several susceptibility loci for NIDDM within the human genome. A putative sugar transporter that has been localized to human chromosome 20q12-q13.1, one of the genomic loci associated with NIDDM. Because of the strong resemblance of this novel protein to members of the mammalian facilitative glucose transporter family (GLUT), the protein is known as GLUT10 (HGMW-approved gene symbol SLC2A10). Data suggests that GLUT10 an excellent candidate for a susceptibility gene involved in NIDDM.
Specifications

Specifications

GLUT10
Polyclonal
Unconjugated
SLC2A10
Liquid
SLC2A10
Synthetic Peptide: I(367) L S T A K K T K P H P R S G D P S A(385)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
O95528
Glucose transporter type 10, Solute carrier family 2, facilitated glucose transporter member 10
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
81031
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit