Learn More
GLUT3, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA121043
Catalog No. PA121043
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Glucose is fundamental to the metabolism of mammalian cells. Several glucose transporter protein (Glut) isoforms have been identified and shown to function in response to insulin and IGF1 induced signaling. GLUT3 is detectable in a few normal cell type spermatids in testis with active spermatogenesis, placental trophoblast membranes, and neurons in brain. GLUT3 staining is also detectable in human cancers including those of the ovary, lung, and testis.
PA1-21043 detects glucose transporter (GLUT3) in human and rat samples.PA1-21043 has been successfully used in immunohistochemistry (paraffin) procedures.The PA1-21043 immunogen is a synthetic peptide to the C-terminal residues of human GLUT3 protein.
Specifications
Specifications
GLUT3
Polyclonal
Unconjugated
SLC2A3
Liquid
SLC2A3
Synthetic peptide to the C-terminal residues of human GLUT3 protein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Rat
Immunohistochemistry (Paraffin), Western Blot, Western Blotting
0.2mg/mL
PBS with 1% BSA and 0.09% sodium azide; pH 7.4
P11169, Q07647
Glucose Transporter 3
Rabbit
IgG
500μL
4° C, do not freeze, do not freeze
Primary
25551, 6515