Learn More
GLUT3 Rabbit anti-Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA185477
Catalog No. PA185477
Add to cart
Description
Description
Rabbit anti Mouse glucose transporter 3 antibody recognizes the mouse glucose transporter 3 (GLUT3), also known as Solute carrier family 2, facilitated glucose transporter member 3 or Glucose transporter type 3, brain.
Glucose is fundamental to the metabolism of mammalian cells. Several glucose transporter protein (Glut) isoforms have been identified and shown to function in response to insulin and IGF-1 induced signaling. GLUT-3 is detectable in a few normal cell type spermatids in testis with active spermatogenesis, placental trophoblast membranes, and neurons in brain. GLUT-3 staining is also detectable in human cancers including those of the ovary, lung, and testis.
Specifications
Specifications
GLUT3
Polyclonal
whole serum with 0.09% sodium azide
P32037, Q07647
FLJ90380, GLUT3
Rabbit
IgG
RUO
Antibody
Polyclonal
Mouse, Rat
Immunoprecipitation, Western Blot
Unconjugated
Slc2a3
Liquid
SLC2A3
13 amino acid synthetic C-terminal sequence of murine GLUT3, conjugated to KLH.
100 μL
4°C or -20°C if preferred
Primary
20527, 25551