Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GluR δ2 Mouse, Unlabeled, Clone: 48, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611006

Catalog No. BDB611006


Add to cart

Description

Description

Glutamate is a major excitatory neurotransmitter of the CNS. The diversity of glutamate is exemplified by two distinct groups of receptors: ionotropic and metabotropic. Ionotroic receptors are ligand-gated cation channels. They can be subdivided into two classes: NMDA and AMPA/kainate receptors. GluR δ2 exhibits only 25% amino acid identity to either ionotropic receptor type. It presents selective and abundant exression in cerebellar Purkinje cells. It is targeted to a subset of Purkinje cell spines and is coexpressed with ionotropic receptors. GluR δ2 is involved in motor coordination, Purkinje cell synapse formation, and cerebellar long-term depression (LTD). The latter is a decrease in the efficacy of the synaptic transmission between parallel fibers and Purkinje neurons and is a cellular basis of motor learning. In fact, mGluR δ2-deficient mice lack LTD. Additionally, an amino acid substitution in transmembrane III of δ2 is responsible for the neurodegeneration seen in Lurcher mice. This substitution is a gain of function mutation that results in disruption of Purkinje membrane potential. Thus, GluR δ2 is an important regulatory component of the Purkinje GluR channel.

Immunofluorescence, Western Blotting

Specifications

Specifications

GluR δ2
48
Unconjugated
Affinity Purified
Mouse GluR δ2 aa. 665-786
50μg
Cell Biology
Primary
Murine, Rabbit, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit