Learn More
GluR2/GluR3, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA137773
Description
The ion channels activated by glutamate are typically divided into two classes. Those that are sensitive to N-methyl-D-aspartate (NMDA) are designated NMDA receptors (NMDAR) while those activated by alpha-amino-3-hydroxy-5 -methyl-4-isoxalone propionic acid (AMPA) are known as AMPA receptors (AMPAR). The AMPAR are comprised of four distinct Glutamate Receptor Subunits designated (GluR1-4) and they play key roles in virtually all excitatory neurotransmission in the brain (Keinänen et al., 1990; Hollmann and Heinemann, 1994). The GluR2 subunit is thought to play a key role in forms of synaptic plasticity such as LTD (Chung et al., 2003).
This antibody is predicted to react with mouse, rat, monkey and avian based on sequence homology.Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is cerebellum tissue.
Specifications
GluR2/GluR3 | |
Polyclonal | |
GRIA3 | |
Glutamate receptor 2, GluR-2, AMPA-selective glutamate receptor 2, GluR-B, GluR-K2, Glutamate receptor ionotropic, AMPA 2, GluA2, GRIA2, GLUR2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2891, 2892 | |
Store at 4°C, do not freeze | |
Liquid |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
P42262, P42263 | |
GRIA2, GRIA3 | |
Synthetic peptide from C-terminus of human GluR2/3 | |
7 mL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.