Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GluR2/GluR3 Rabbit anti-Avian, Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA137774

Catalog No. 01-673-513


Add to cart

Description

Description

This antibody is predicted to react with mouse, rat, non-human primate, and avian based on sequence homology. Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is cerebellum tissue.

Glutamate receptors constitute the principal excitatory neurotransmitter receptors in brain. Two classes of glutamate receptors exist: Ionotropic receptors, and metabotropic receptors. Ionotropic glutamate receptors are oligomeric complexes of various subunits (GluR1-7, NMDA1-3, KAI-2), which comprise ligand-gated calcium channels. Metabotropic glutamate receptors are G-protein coupled receptors, which when activated can activate phopholipases or adenylylcyclase, depending on the neuron.
Specifications

Specifications

GluR2/GluR3
Polyclonal
PBS with 1% BSA and 0.1% sodium azide; pH 7.4
P42262, P42263
Glutamate receptor 2, GluR-2, AMPA-selective glutamate receptor 2, GluR-B, GluR-K2, Glutamate receptor ionotropic, AMPA 2, GluA2, GRIA2, GLUR2
Rabbit
IgG
1 mL
-20° C, Avoid Freeze/Thaw Cycles
Primary
2891, 2892
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
GRIA3
Liquid
GRIA2, GRIA3
Synthetic peptide from C-terminus of human GluR2/3
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Avian, Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit