Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GluR6/GluR7 Rabbit anti-Human, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA137780

Catalog No. 01-673-519


Add to cart

Description

Description

This antibody is predicted to react with bovine, chicken, canine, amphibian, mouse, rabbit and rat based on sequence homology. Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is brain tissue.

Glutamate receptors constitute the principal excitatory neurotransmitter receptors in brain. Two classes of glutamate receptors exist: ionotropic receptors, and metabotropic receptors. Ionotropic glutamate receptors are oligomeric complexes of various subunits (GluR1-7, NMDA1-3, KAI-2), which comprise ligand-gated calcium channels. Metabotropic glutamate receptors are G-protein coupled receptors, which when activated can activate phopholipases or adenylylcyclase, depending on the neuron.
Specifications

Specifications

GluR6/GluR7
Polyclonal
PBS with 1% BSA and <0.1% sodium azide; pH 7.6
P42260, Q13002, Q13003
GRIK2, GRIK3, EAA4, GLR6, MRT6, GLUK6, GLUR6, GluK2, Glutamate Receptor 6/7, Glutamate Receptor 6, Glutamate Receptor 7
Rabbit
IgG
1 mL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
54257, 2898, 2899
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
GRIK3
Liquid
GRIK2, GRIK3
Synthetic peptide from C-terminus of human GluR6/7.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit