Learn More
GLUT3 Rabbit anti-Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OSG00012W
Catalog No. PIOSG00012W
Add to cart
Description
Description
Reconstitute with 100 µL of distilled water.
Specifications
Specifications
GLUT3
Polyclonal
whole serum with no preservative
P32037, Q07647
GLUT-3, GLUT3, Slc2a3, Solute carrier family 2, facilitated glucose transporter member 3
Rabbit
IgG
RUO
Antibody
Polyclonal
Mouse, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Unconjugated
Slc2a3
Lyophilized
SLC2A3
A synthetic peptide from aa region 450-493 of rat GLUT3 conjugated to an immunogenic carrier protein was used as the antigen
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
20527, 25551