Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLUT3, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA128204

Catalog No. PA128204

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PA1-28204 detects GLUT3 from human samples.PA1-28204 has been successfully used in immunohistochemistry (paraffin tissue) applications.The PA1-28204 immunogen is: Synthetic peptide (unfortunately, the amino acid sequence is considered to be commercially sensitive) (Human) (C terminal).
Specifications

Specifications

GLUT3
Polyclonal
PBS with 1% BSA and 0.1% sodium azide; pH 7.4
P11169, Q07647
FLJ90380, GLUT3
Rabbit
IgG
7mL
Store at 4°C, do not freeze
Primary
6515
Immunohistochemistry (Paraffin)
Unconjugated
SLC2A3
Purified
SLC2A3
Synthetic peptidecorresponding to the Human C terminal region.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit