Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glut4 Rabbit anti-Human, Mouse, Rat, Plant, Biotin, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP149533B

Catalog No. NBP149533B


Add to cart

Description

Description

Glut4 Polyclonal antibody specifically detects Glut4 in Human, Mouse, Rat, Plant samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown.
Specifications

Specifications

Glut4
Biotin
PBS with 0.05% Sodium Azide
Affinity Purified
SLC2A4
A synthetic peptide made to a C-terminal portion of the human Glucose Transporter GLUT4 protein (between residues 480-509) [UniProt P14672]
Immunogen affinity purified
RUO
Store at 4C in the dark.
Polyclonal
Human and mouse. Predicted to react with rat based on 100% sequence homology.
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown, Western Blot
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, KnockDown Validated
P14672
GLUT-4, GLUT4Glucose transporter type 4, insulin-responsive, insulin-responsive glucose transporter type 4, solute carrier family 2 (facilitated glucose transporter), member 4, solute carrier family 2, facilitated glucose transporter member 4
Rabbit
IgG
0.1 ml
Diabetes Research, Lipid and Metabolism, Membrane Trafficking and Chaperones
Primary
6517
Human, Mouse, Plant, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only