Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-GLUT9, Alexa Fluor 405, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP106565AF405
Description
GLUT9 Polyclonal specifically detects GLUT9 in Human, Mouse, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
GLUT9 | |
Polyclonal | |
Western Blot, Immunohistochemistry-Paraffin | |
Q9NRM0 | |
SLC2A9 | |
Synthetic peptide made to an internal portion of human GLUT9 (within residues 10-60). [Swiss-Prot# Q9NRM0] | |
Affinity Purified | |
RUO | |
Primary | |
This antibody is specific for the human GLUT9 protein Isoform 1 (SLC2A9-L). | |
Store at 4C in the dark. |
Western Blot, Immunohistochemistry (Paraffin) | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
facilitated glucose transporter member 9, GLUT-9, GLUT9Glut9, GLUTX, solute carrier family 2 (facilitated glucose transporter), member 9, UAQTL2, urate voltage-driven efflux transporter 1 | |
Rabbit | |
58.7 kDa | |
0.1 mL | |
Lipid and Metabolism, Plasma Membrane Markers, Signal Transduction | |
56606 | |
Human, Mouse, Primate | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only