Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutamine Synthetase Mouse, Unlabeled, Clone: 6, BD

Mouse Monoclonal Antibody

$232.00 - $458.00

Specifications

Antigen Glutamine Synthetase
Isotype IgG2a
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB610517
View Documents
BD Biosciences
610517
50μg
Each for $232.00
Add to cart
 
BDB610518
View Documents
BD Biosciences
610518
150μg
Each for $458.00
Add to cart
 
Description

Description

Glutamine synthetase catalyzes the amination of glutamic acid to form glutamine. It is found in mammals as an octamer of identical 45 kDa subunits. Glutamine synthetase activity is a useful marker for astrocytes and an important differentiation feature in retina. It is also considered to be a key enzyme in the recycling of the neurotransmitter glutamate.

Specifications

Specifications

Glutamine Synthetase
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Unconjugated
Cell Biology
Mouse
Affinity Purified
Monoclonal
Glutamate-Ammonia Ligase; GLUL; GLNS
IgG2a
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Primary
Western Blot
RUO
6
250μg/mL
Store undiluted at -20°C.
SDS
Product Certifications

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit