Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Glutathione Peroxidase 1/GPX1, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 5 publications

Manufacturer:  Novus Biologicals NBP133620

Catalog No. NBP133620


Add to cart

Description

Description

Glutathione Peroxidase 1/GPX1 Polyclonal specifically detects Glutathione Peroxidase 1/GPX1 in Human, Mouse, Rat, Alligator samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunoblotting.
Specifications

Specifications

Glutathione Peroxidase 1/GPX1
1.0mg/mL
Western Blot 1:1000-1:10000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000, Immunoblotting
Affinity Purified
GPX1
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human GPX1. The exact sequence is proprietary.
22 kDa
100 ul
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Expected to cross-reactivity based on sequence identity: Rhesus Monkey (100%) and Chimpanzee (100%).
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
PBS, 20% Glycerol (pH7). with 0.025% Proclin 300
Cellular glutathione peroxidase, EC 1.11.1, EC 1.11.1.9, glutathione peroxidase 1, GPx-1, GSHPx-1, GSHPX1, MGC14399, MGC88245
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
2876
Human, Mouse, Rat
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only