Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GSTM1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA527153
Description
Recommended positive controls: HeLa. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
GSTM1 Polyclonal antibody specifically detects GSTM1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
GSTM1 | |
Polyclonal | |
Unconjugated | |
GSTM1 | |
glutathione S-alkyltransferase; glutathione S-aralkyltransferase; glutathione S-aryltransferase; glutathione S-transferase GT8.7; glutathione S-transferase M1; glutathione S-transferase Mu 1; Glutathione S-transferase Yb-1; glutathione S-transferase Yb-1 subunit; glutathione S-transferase, mu 1; glutathione-S-transferase mu type 1 (Yb1); glutathione-S-transferase, mu 1; glutathione-S-transferase, mu type 1 (Yb1); GST 1-1; GST 3-3; GST class-mu 1; GST HB subunit 4; GST Yb1; GST1; GSTA3; Gstb1; Gstb-1; GSTM1; GSTM1-1; GSTM1a-1a; GSTM1b-1b; GTH4; GTM1; H-B; HB subunit 4; MU; MU-1; pmGT10; S-(hydroxyalkyl)glutathione lyase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2944 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
P09488 | |
GSTM1 | |
Synthetic peptide corresponding to a region within amino acids 158 and 168 of Human GSTM1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction