Learn More
MilliporeSigma™ anti-Glutathione S-Transferase, Polyclonal
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma™ PC53850UL
Catalog No. PC53850UL
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Specifically detects Glutathione S-Transferase Clone: in S. japonicum samples, and it is validated for Affinity Purification, ELISA, Immunoprecipitation, Immuno Blotting
Glutathione S-transferase (GST) is commonly used as an epitope tag in generating fusion proteins. In these systems a~26kDa GST domain isolated fromS. japonicum is cloned in-frame to the gene encoding the protein of interest in the appropriate expression vector system. The resulting fusion protein can then be expressed by eitherE. coli (pGEX expression vector system) or by yeast (pYEX™ expression vector system) in a soluble form. The resulting fusion proteins may then be further purified by immunoprecipitation or affinity chromatography with GST (Ab-1) or directly analyzed by western blotting with the antibody.
Specifications
Specifications
Glutathione S-Transferase
Unconjugated
purified
recombinant glutathione S-transferase from S. japonicum
50μL
2°C to 8°C
Polyclonal
S. japonicum
Affinity Purification, ELISA, Immuno Blotting, Immunoblot, Immunoprecipitation
In PBS, 0.2% Tween™-20 detergent.
Rabbit
IgG
RUO
Primary
Recognizes glutathione S-transferase. Good negative control for rabbit polyclonal antibodies.