Learn More
Glycerol kinase 2 Mouse anti-Human, Clone: 3G4, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA518967
Catalog No. PIMA518967
Add to cart
Description
Description
Peptide Sequence: MAAPKTAAVG PLVGAVVQGT NSTRFLVFNS KTAELLSHHK VELTQEFPKE GWVEQDPKEI LQSVYECIAR TCEKLDELNI DISNIKAVGV SNQRETTVIW DKLTGEPLYN AVVWLDLRTQ TTVEDLSKKI PGNSNFVKSK TGLPLSTYFS AVKLRWMLDN VRNVQKAVEE GRALFGTIDS WLIWSLTGGV NGGVHCTDVT NASRTMLFNI HSLEWDKELC DFFEIPMDLL PNVFSSSEIY GLIKTGALEG VPISGCLGDQ CAALVGQMCF QEGQAKNTYG TGCFLLCNTG RKCVFSEHGL LTTVAYKLGR EKPAYYALEG SVAIAGAVIR WLRDNLGIIE TSGDIERLAK EVGTSYGCYF VPAFSGLYAP YWEPSARGIL CGLTQFTNKC HIAFAALEAV CFQTREILEA MNRDCGIPLR HLQVDGGMTN NKVLMQLQAD ILHIPVIKPF MPETTALGAA MAAGAAEGVS VWSLEPQALS VLRMERFEPQ IQATESEIRY ATWKKAVMKS MGWVTSQSPE GGDPSIFSSL PLGFFIVSSM VMLIGARYIS GVP
The human glycerol kinase gene family consists of at least 3 expressed loci. The GK1 locus on Xp21.3 is the site of mutations (deletions) causing glycerol kinase deficiency. It comprises 19 exons and is probably ancestral to several other genes which, because they are intronless, are suspected of having arisen by reverse transcriptase mediated events. These include 2 genes on chromosome 4. They are expressed as a single mRNA species in testis where expression is at a high level.
Specifications
Specifications
Glycerol kinase 2
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
Q14410
GKP2, GKTA, ATP:glycerol 3-phosphotransferase 2, GK 2, glycerokinase 2, glycerol kinase pseudogene 2, glycerol kinase testis specific 2, glycerol kinase, testis specific 2
Mouse
IgG2a, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
2712
ELISA, Western Blot
3G4
Unconjugated
GK2
Liquid
GK2
GK2 (AAH29820, 1 a.a. ∽ 553 a.a) full-length recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human