Learn More
Glycine Binding Protein Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA3100
Catalog No. PA3100
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
PA3-100 detects the glycine-binding protein. PA3-100 has been used successfully in Western blot, ELISA, immunohistochemistry, immunocytochemistry, and immunoprecipitation procedures. In Western blot analysis of rat brain synaptic membranes this antibody detects a ~60 kDa protein representing glycine-binding protein. The PA3-100 immunogen is a synthetic peptide corresponding to the glycine-binding region residues S(140) Q V D L L A R L L R D L G(153) C of rat GBP.
Ionotropic glutamate receptors are oligomeric ion channel proteins, which mediate fast neurotransmission and excitatory synapses in the mammalian central nervous system. There are three distinct subfamilies, of which NMDA receptors have received particular attention because they are implicated in both synaptic plasticity and the pathogenesis of acute and chronic neurodegenerative disorders. A unique feature of the NMDA receptors is their requirement for both glutamate and the co-agonist glycine for efficient gating. Glycine increases the affinity of the receptor for the endogenous agonist glutamate. The glycine binding site is found on the NR1 subunit of the NMDA receptor.
Specifications
Specifications
Glycine Binding Protein
Polyclonal
whole serum with 0.05% sodium azide
O60308, Q80V31, D3Z8X7
CEP104
Synthetic peptide corresponding to the glycine-binding region residues S(140) Q V D L L A R L L R D L G(153) C of rat GBP.
200 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
230967, 246295, 9731
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Unconjugated
CEP104
Liquid
Rabbit
IgG
RUO
Antibody
Polyclonal
Human, Mouse, Rat