Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glycine Receptor α3 Subunit Rabbit anti-Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
$495.00 - $927.00
Specifications
Antigen | Glycine Receptor α3 Subunit |
---|---|
Applications | Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AB547250UL
|
MilliporeSigma
AB547250UL |
50 μL |
Each for $495.00
|
|
|||||
AB5472200UL
|
MilliporeSigma
AB5472200UL |
200 μL |
Each for $927.00
|
|
|||||
Description
This Anti-Glycine Receptor α3 Subunit Antibody is validated for use in immunohistochemistry for the detection of Glycine Receptor α3 Subunit.Specifications
Glycine Receptor α3 Subunit | |
Polyclonal | |
Rabbit | |
NM_001042543.1; NM_006529.2 | |
A purified peptide corresponding to amino acid residues 355-368 of rat GlyRa3 (Accession P24524). | |
Affinity Purified |
Immunohistochemistry | |
Unconjugated | |
Mouse, Rat | |
GLRA3 | |
Primary | |
−20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title