Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GLRA1 Monoclonal Antibody (4D6), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA518977
Description
Peptide Sequence: IWKPDLFFAN EKGAHFHEIT TDNKLLRISR NGNVLYSIRI TLTLACPMDL KNFPMDVQTC IMQLESFGYT MNDLIFEWQE QGAVQVADGL TLPQFILKEE
GLRA1 is a subunit of a pentameric inhibitory glycine receptor, which mediates postsynaptic inhibition in the central nervous system. Defects in GLRA1 are a cause of startle disease (STHE), also known as hereditary hyperekplexia or congenital stiff-person syndrome. Multiple transcript variants encoding different isoforms have been found for GLRA1. Glycine receptors (GlyR) that include GLRA1 are members of the cys-loop family of ligand-gated ion channels, responsible for mediating the inhibitory effects of glycine. GLRA1 are widely distributed throughout the CNS, particularly within the hippocampus, spinal cord and brain stem. Diseases associated with GLRA1 include Hyperekplexia, Hereditary 1 and Hyperekplexia.Specifications
GLRA1 | |
Monoclonal | |
0.2-1.0 mg/mL | |
PBS with no preservative; pH 7.4 | |
P23415 | |
GLRA1 | |
GLRA1 (NP_000162, 121 a.a. ∽ 220 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Western Blot | |
4D6 | |
Unconjugated | |
GLRA1 | |
GLRA1, Glra2, GLYRA1, Glycine receptor subunit alpha-1, Glycine receptor subunit alpha-2 | |
Mouse | |
Affinity chromatography | |
RUO | |
2741 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction