Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycogen Phosphorylase BB/GPBB Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP132799

Catalog No. NBP132799


Add to cart

Description

Description

Glycogen Phosphorylase BB/GPBB Polyclonal antibody specifically detects Glycogen Phosphorylase BB/GPBB in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

Glycogen Phosphorylase BB/GPBB
0.84mg/mL
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
P11216
Brain glycogen phosphorylase, EC 2.4.1.1, Glycogen phosphorylase B, Glycogen phosphorylase brain form, Glycogen Phosphorylase Isoenzyme BB, MGC9213, Phosphorylase glycogen brain, PYGB
Rabbit
IgG
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse, Rat
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Affinity Purified
PYGB
Recombinant protein encompassing a sequence within the C-terminus region of human GPBB. The exact sequence is proprietary.
97 kDa
0.1 ml
GPCR
Primary
5834
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only