Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD235a Mouse anti-Human, Clone: A84-B/H2, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA135460

Catalog No. 01-671-313

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Glycophorins A (GPA) and B (GPB) which are single pass membrane sialoglycoproteins. GPA is the carrier of blood group M and N specificities, while GPB accounts for S and U specificities. GPA and GPB provide the cells with a large mucin like surface and it has been suggested this provides a barrier to cell fusion, so minimizing aggregation between red blood cells in the circulation.

Specifications

Specifications

CD235a
Monoclonal
100 μg/mL
PBS with 0.05% sodium azide
P02724
GYPA, glycophorin A, HGNC:4702, GPA, GPErik, GPSAT, HGpMiV, HGpMiX, HGpMiXI, HGpStaC, MN, MNS , Mi.V glycoprotein
Mouse
IgG2a
100 μg
4° C
Primary
2993
ELISA, Immunohistochemistry (Frozen)
A84-B/H2
Unconjugated
GYPA
Liquid
GYPA
Human erythrocytes, treated with neuraminidase.
Purified
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit