Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P-Glycoprotein Mouse anti-Hamster, Human, Clone: JSB-1, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA135604

Catalog No. 01-671-042

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MDR/TAP subfamily. Members of the MDR/TAP subfamily are involved in multidrug resistance. The protein encoded by this gene is an ATP-dependent drug efflux pump for xenobiotic compounds with broad substrate specificity. It is responsible for decreased drug accumulation in multidrug-resistant cells and often mediates the development of resistance to anticancer drugs. This protein also functions as a transporter in the blood-brain barrier.

This antibody reacts with a conserved cytoplasmic epitope of the plasma membrane-associated 170-180 kD glycoprotein, the expression of which is strongly correlated with the degree of multi-drug-resistance (MDR) derived cell lines, including cell lines derived from lung, ovaries and B cell lymphomas. This product cross-reacts with chinese hamster but does not cross-react with mouse or rat.
Specifications

Specifications

P-Glycoprotein
Monoclonal
Unconjugated
tissue culture supernatant with 1% BSA and 0.1% sodium azide
P08183
ABC20, CD243, CLCS, GP170, MDR1, P-GP, PGY1, P-glycoprotein 1, colchicin sensitivity, doxorubicin resistance, multidrug resistance protein 1, CD243
Mouse
IgG1
RUO
Antibody
Monoclonal
Hamster, Human
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
JSB-1
Assay dependent
ABCB1
Liquid
Abcb1
Multidrug-resistant Chinese hamster ovary cell line (CHrC5).
1 mL
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Primary
100682536, 5243
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit