Learn More
GLYCTK Mouse anti-Human, Mouse, Clone: 1B5, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA523022
Catalog No. PIMA523022
Add to cart
Description
Description
Peptide Sequence: VLSAAMQGDV KSMAQFYGLL AHVARTRLTP SMAGASVEED AQLHELAAEL QIPDLQLEEA LETMAWGRGP VCLLAGGEPT VQLQGSGRGG RNQELALRV
This locus encodes a member of the glycerate kinase type-2 family. The encoded enzyme catalyzes the phosphorylation of (R)-glycerate and may be involved in serine degradation and fructose metabolism. Decreased activity of the encoded enzyme may be associated with the disease D-glyceric aciduria. Alternatively spliced transcript variants have been described.
Specifications
Specifications
GLYCTK
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
Q8IVS8, Q8QZY2
LP5910, HBEBP2, HBEBP4, HBeAgBP4A, HBeAg binding protein 4, HBeAg-binding protein 2
Mouse
IgG1, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
132158, 235582
ELISA, Western Blot
1B5
Unconjugated
GLYCTK
Liquid
GLYCTK
GLYCTK (NP_660305.2, 332 a.a. ∽ 430 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human, Mouse