Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-Glyoxalase I, DyLight 350, Clone: Glo1a, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP119015UV
Description
Glyoxalase I Monoclonal antibody specifically detects Glyoxalase I in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Glyoxalase I | |
Glo1a | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
Purified | |
GLO1 | |
Full length human GLO1 protein [Swiss-Prot #Q04760]. | |
21 kDa | |
0.1 ml | |
Store at 4C in the dark. | |
Monoclonal | |
Human, Mouse |
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Aldoketomutase, EC 4.4.1.5, GLOD1, Glx I, GLYI, glyoxalase domain containing 1, glyoxalase Ialdoketomutase, Ketone-aldehyde mutase, lactoyl glutathione lyase, lactoylglutathione lyase, methylglyoxalase, S-D-lactoylglutathione methylglyoxal lyase | |
Mouse | |
IgG1 Kappa | |
Protein G purified | |
Cancer, Core ESC Like Genes, Signal Transduction, Stem Cell Markers | |
Primary | |
2739 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title