Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glypican 6 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA513387

Catalog No. PIPA513387

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The glypicans comprise a family of glycosylphosphatidylinositol-anchored heparan sulfate proteoglycans. The glypicans have been implicated in the control of cell growth and division. Glypican 6 is a putative cell surface coreceptor for growth factors, extracellular matrix proteins, proteases and anti-proteases.
Specifications

Specifications

Glypican 6
Polyclonal
PBS with 0.09% sodium azide
Q9Y625
Glypican-6 [Precursor]; Secreted glypican-6; GPC6
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10082
Western Blot
Unconjugated
GPC6
Liquid
GPC6
KLH conjugated synthetic peptide between 16-46 amino acids from the N-terminal region of human Glypican 6 (GPC6)
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit