Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GM-CSF Rat anti-Mouse, Brilliant Violet 421, Clone: MP1-22E9, BD

Rat Monoclonal Antibody

Manufacturer:  BD Biosciences 564747

Catalog No. BDB564747


Add to cart

Description

Description

The MP1-22E9 monoclonal antibody specifically binds to mouse Granulocyte-Macrophage Colony Stimulating Factor (GM-CSF). The immunogen used to generate the MP1-22E9 hybridoma was yeast-expressed recombinant mouse GM-CSF. This is a neutralizing antibody.

The antibody was conjugated to BD Horizon BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates.

Specifications

Specifications

GM-CSF
MP1-22E9
Brilliant Violet 421
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Rat
IgG2a
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
0.2mg/mL
Csf2; Csfgm; Colony stimulating factor 2 (granulocyte-macrophage)
Affinity Purified
Recombinant Mouse GM-CSF
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
RUO
Primary
Murine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit