Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GM-CSF Mouse anti-Human, Clone: 3209, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.

Mouse Monoclonal Antibody

$135.50 - $839.00

Specifications

Antigen GM-CSF
Immunogen E. coli-derived recombinant human GM-CSF Ala18-Glu144 Accession # P04141
Clone 3209
Applications Western Blot, Neutralization, Immunocytochemistry
Conjugate Unconjugated
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB215SP
View Documents
R&D Systems
MAB215SP
25 ug
Each for $135.50
Add to cart
 
MAB215100
View Documents
R&D Systems
MAB215100
100 ug
Each for $387.50
Add to cart
 
MAB215500 Available on GSA/VA Contract for Federal Government customers only.
View Documents
R&D Systems
MAB215500
500 ug
Each for $839.00
Add to cart
 
Description

Description

GM-CSF Monoclonal antibody MAB215 Specifically detects GM-CSF in Human samples. It is validated for Immunocytochemistry, Neutralization, Western Blotting
Specifications

Specifications

GM-CSF
3209
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Mouse
IgG1
1437
RUO
E. coli-derived recombinant human GM-CSF Ala18-Glu144 Accession # P04141
Western Blot, Neutralization, Immunocytochemistry
Purified
P04141
Monoclonal
Primary
Detects human GM-CSF in Western blots.
Protein A or G purified from hybridoma culture supernatant
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit