Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GM2A Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525737

Catalog No. PIPA525737


Add to cart

Description

Description

This antibody is predicted to react with non-human primate based on sequence homology.

Specifications

Specifications

GM2A
Polyclonal
PBS with 0.09% sodium azide
P17900, Q60648
GM2-AP, SAP-3, cerebroside sulfate activator protein, ganglioside GM2 activator, shingolipid activator protein 3, sphingolipid activator protein 3
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
14667, 2760
Western Blot
Unconjugated
GM2A
Liquid
GM2A
KLH conjugated synthetic peptide between 25-54 amino acids from the N-terminal region of human GM2A
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit