Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-GMDS, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP133424

Catalog No. NBP133424


Add to cart

Description

Description

GMDS Polyclonal antibody specifically detects GMDS in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

GMDS
1.0mg/mL
Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000
O60547
EC 4.2.1.47, GDP-D-mannose dehydratase, GDP-mannose 4,6-dehydratase, GMDGDP-mannose 4,6 dehydratase, SDR3E1, short chain dehydrogenase/reductase family 3E, member 1
Rabbit
IgG
100ul
Stem Cell Markers
Primary
2762
Human
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Affinity Purified
GMDS
Recombinant protein encompassing a sequence within the center region of human GMDS. The exact sequence is proprietary.
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Predicted cross-reactivity based on sequence identity: Xenopus laevis (93%), Zebrafish (89%).
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only