Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-GMPS, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP216696
Description
GMPS Polyclonal specifically detects GMPS in Human samples. It is validated for Western Blot.Specifications
GMPS | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
EC 6.3.5.2, Glutamine amidotransferase, GMP synthase, GMP synthase [glutamine-hydrolyzing], GMP synthetase, guanine monphosphate synthetase, guanosine 5'-monophosphate synthase, MLL/GMPS fusion protein | |
Rabbit | |
77 kDa | |
0.1 mL | |
metabolism | |
8833 | |
Human | |
IgG |
Western Blot | |
0.69mg/mL | |
Western Blot 1:500-1:3000 | |
P49915 | |
GMPS | |
Recombinant protein encompassing a sequence within the center region of human GMP synthase. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Human. Expected to cross react based on sequence identity: Mouse (96%), Chicken (91%), Rat (97%), Zebrafish (85%), Bovine (97%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only