Learn More
MilliporeSigma™ anti-GNA13 Clone: 6F6-B5, Available on GSA/VA Contract for Federal Government customers only.
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma™ ST1629100UG
Catalog No. ST1629100UG
Add to cart
Description
Description
Specifically detects GNA13 Clone: 6F6-B5 in Human samples, and it is validated for ELISA, Immuno Blotting
Heterotrimeric G proteins are membrane bound GTPases that are linked to 7-TM receptors. Each G protein contains an α-, β- and γ-subunit and is bound to GDP in the 'off' state. Ligand binding to a 7-TM receptor causes a conformational change in the receptor, which detaches the G protein. This reaction switches the G protein to an 'on' state, allowing GTP to displace GDP, thus Gα can activate second messenger signaling cascades. There are several types of Gα proteins; Gαs and Gαi bind directly to adenylyl cyclase and stimulate or inhibit its activity, leading to an increase or decrease in cAMP levels respectively. Gαq/11 stimulates PLCβ which catalyzes the hydroylsis of PIP2 to the second messengers IP3 and DAG. Gv12/13 are involved in Rho family GTPase signaling. In addition, some Gβγ subunits have active functions; Gβv coupled to H1 receptors can activate PLA2 and Gβγ coupled to M1 receptors can activate KIR channels. Rapid termination of the cellular response is required once the concentration of the stimulating ligand decreases. GTP bound to Gα is hydrolyzed rapidly to GDP, Gα is converted to its inactive form and re-associates with the β- and γ-subunits returning to the 'off' state.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
GNA13
6F6-B5
In PBS, pH 7.2.
Murine
IgG1
RUO
Primary
Recognizes the ∽40-45kDa GNA13 protein in HepG2 cells.
ELISA, Immuno Blotting, Immunoblot
Unconjugated
Purified
Full-length
100μg
−20°C
Monoclonal
Human