Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

guanine nucleotide binding protein (G protein), alpha activating activity polypeptide, olfactory type, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a partial recombinant GNAL.

Manufacturer:  Abnova Corporation H00002774A01

Catalog No. 89-004-138


Add to cart

Description

Description

Sequence: DMLAEKVLAGKSKIEDYFPEYANYTVPEDATPDAGEDPKVTRAKFFIRDLFLRISTATGDGKHYCYPHFTCAVDTENIRRVFNDCRDIIQRMHLKQYELL
Specifications

Specifications

guanine nucleotide binding protein (G protein), alpha activating activity polypeptide, olfactory type
Unconjugated
50% glycerol
NM_182978
GNAL
GNAL (NP_892023, 359 a.a. ~ 458 a.a) partial recombinant protein with GST tag.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
2774
ELISA, Western Blot
Mouse polyclonal antibody raised against a partial recombinant GNAL.
GNAL
Serum
Mouse
50 uL
Signal Transduction
Y
Polyclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit