Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

guanine nucleotide binding protein (G protein), alpha z polypeptide, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human GNAZ protein.

Manufacturer:  Abnova Corporation H00002781B01

Catalog No. 89-005-454


Add to cart

Description

Description

The protein encoded by this gene is a member of a G protein subfamily that mediates signal transduction in pertussis toxin-insensitive systms. This encoded protein may play a role in maintaining the ionic balance of perilymphatic and endolymphatic cochlear fluids. [provided by RefSeq

Sequence: MGCRQSSEEKEAARRSRRIDRHLRSESQRQRREIKLLLLGTSNSGKSTIVKQMKIIHSGGFNLEACKEYKPLIIYNAIDSLTRIIRALAALRIDFHNPDRAYDAVQLFALTGPAESKGEITPELLGVMRRLWADQGAQACFSRSSEYHLEDNAAYYLNDLERIAAADYIPTVEDILRSRDMTTGIVENKFTFKELTFKMVDVGGQRSERKKWIHCFEGVTAIIFCVELSGYDLKLYEDNQTSRMAESLRLFDSICNNNWFINTSLILFLNKKDLLAEKIRRIPLTICFPEYKGQNTYEEAAVYIQRQFEDLNRNKETKEIYSHFTCATDTSNIQFVFDAVTDVIIQNNLKYIGLC
Specifications

Specifications

guanine nucleotide binding protein (G protein), alpha z polypeptide
Unconjugated
No additive
BC026342
GNAZ
GNAZ (AAH26342, 1 a.a. ~ 355 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
2781
Western Blot
Mouse polyclonal antibody raised against a full-length human GNAZ protein.
GNAZ
Serum
Mouse
50 uL
Membrane Receptors
Y
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit