Learn More
anti-GNB1, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232757
Catalog No. NBP232757
Add to cart
Description
Description
GNB1 Polyclonal antibody specifically detects GNB1 in Zebrafish samples. It is validated for Western Blot.
Specifications
Specifications
GNB1
Unconjugated
PBS with 0.09% Sodium Azide
Affinity Purified
GNB1
This DANRE gnb1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of DANRE gnb1.
37.286 kDa
400 ul
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Zebrafish
Western Blot
Western Blot 1:1000
Q6PH57
beta subunit, signal-transducing proteins GS/GI, G protein, beta-1 subunit, guanine nucleotide binding protein (G protein), beta polypeptide 1, guanine nucleotide-binding protein G(I)/G(S)/G(T) beta subunit 1, guanine nucleotide-binding protein G(I)/G(S)/G(T) subunit beta-1, Transducin beta chain 1
Rabbit
IgG
Protein A purified
RUO
Primary
2782