Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GNE Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525238

Catalog No. PIPA525238


Add to cart

Description

Description

This antibody is predicted to react with hamster and rat based on sequence homology.

GNE is a bifunctional enzyme that initiates and regulates the biosynthesis of N-acetylneuraminic acid (NeuAc), a precursor of sialic acids. It is a rate-limiting enzyme in the sialic acid biosynthetic pathway. Sialic acid modification of cell surface molecules is crucial for their function in many biologic processes, including cell adhesion and signal transduction. Differential sialylation of cell surface molecules is also implicated in the tumorigenicity and metastatic behavior of maligt cells.
Specifications

Specifications

GNE
Polyclonal
PBS with 0.09% sodium azide
Q9Y223, Q91WG8
DMRV, GLCNE, IBM2, NM, Uae1, N-acylmannosamine kinase, UDP-GlcNAc-2-epimerase/ManAc kinase, UDP-N-acetylglucosamine 2-epimerase/N-acetylmannosamine kinase, UDP-N-acetylglucosamine-2-epimerase/N-acetylmannosamine kinase, bifunctional UDP-N-acetylglucosamine 2-epimerase/N-acetylmannosamine
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10020, 50798
Flow Cytometry, Western Blot
Unconjugated
GNE
Liquid
GNE
KLH conjugated synthetic peptide between 580-607 amino acids from the C-terminal region of human GNE
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit