Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

guanine nucleotide binding protein-like 1, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human GNL1 protein.

Manufacturer:  Abnova Corporation H00002794B01

Catalog No. 89-005-457


Add to cart

Description

Description

The GNL1 gene, identified in the human major histocompatibility complex class I region, shows a high degree of similarity with its mouse counterpart. The GNL1 gene is located less than 2kb centromeric to HLA-E, in the same transcriptional orientation. GNL1 is telomeric to HLA-B and HLA-C. [provided by RefSeq

Sequence: MPRKKPFSVKQKKKQLQDKRERKRGLQDGLRSSSNSRSGSRERREEQTDTSDGESVTHHIRRLNQQPSQGLGPRGYDPNRYRLHFERDSREEVERRKRAAREQVLQPVSAELLELDIREVYQPGSVLDFPRRPPWSYEMSKEQLMSQEERSFQDYLGKIHGAYSSEKLSYFEHNLETWRQLWRVLEMSDIVLLITDIRHPVVNFPPALYEYVTGELGLALVLVLNKVDLAPPALVVAWKHYFHQHYPQLHVVLFTSFPRDPRTPQDPSSVLKKSRRRGRGWTRALGPEQLLRACEAITVGKVDLSSWREKIARDVAGATWGNGSGEEEEEEDGPAVLVEQQTDSAMEPTGPTQERYKDGVVTIGCVGFPNVGKSSLINGLVGRKVVSVSRTPGHTRYFQTYFLTPSVKLCDCPGLIFPSLLPRQLQVLAGIYPIAQIQEPYTAVGYLASRIPVQALLHLRHPEAEDPSAEHPWCAWDICEAWAEKRGYKTAKAARNDVYRAANSLLRLAVDGRLSLCFHPPGYSEQKGTWESHPETTELVVLQGRVGPAGDEEEEEEEELSSSCEEEGEEDRDADEEGEGDEETPTSAPGSSLAGRNPYALLGEDEC
Specifications

Specifications

guanine nucleotide binding protein-like 1
Unconjugated
No additive
NM_005275.2
HSR1
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
2794
Immunofluorescence, Western Blot
Mouse polyclonal antibody raised against a full-length human GNL1 protein.
GNL1
Serum
GNL1
GNL1 (NP_005266.2, 1 a.a. ~ 607 a.a) full-length human protein.
RUO
Y
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit