Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GNPDA1 Rabbit anti-Equine, Human, Non-human primate, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523241

Catalog No. PIPA523241


Add to cart

Description

Description

GNPDA1 is a 289 amino acid cytoplasmic protein belonging to the glucosamine/ galactosamine-6-phosphate isomerase family. It is one of the two mammalian glucosamine-6-phosphate deaminase enzymes. GNPDA1 is an allosteric enzyme that catalyzes the reversible conversion of D-glucosamine-6-phosphate (GlcN6P) into D-fructose-6-phosphate (Fru6P) and ammonium with an aldo/keto isomerization and an amination/deamination. GNPDA1 may trigger calcium oscillations in mammalian eggs at fertilization which in turn serve as an important factor for egg development and early embryo development. It is a homohexamer, ubiquitously expressed in most of human tissues with high expression in testis, ovary, placenta, and heart and also in various cell lines.
Specifications

Specifications

GNPDA1
Polyclonal
Unconjugated
GNPDA1
Liquid
GNPDA1
Synthetic peptide (266 VDPLYSIKEKETEKSQSS 283) of GNPDA1 protein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Equine, Human, Non-human Primate
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
P46926
GNP1, GNPDA, GNPI, GPI, HLN, GNPDA 1, glcN6P deaminase 1, glucosamine-6-phosphate isomerase 1, oscillin
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
100061298, 10007
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit