Learn More
Thermo Scientific Lab Vision GnRH Receptor/LH-RH Receptor Ab-3, Mouse Monoclonal Antibody
Obtain accurate, reproducible results in immunohistochemistry, immunofluorescence and flow cytometry experiments.
Manufacturer: Thermo Scientific MS1139P1
Description
Obtain accurate, reproducible results in immunohistochemistry, immunofluorescence and flow cytometry experiments with Thermo Scientific GnRH Receptor/LH-RH Receptor Ab-3, Mouse Monoclonal Antibody.
Host Species: Mouse
Clone: GNRH03; same as F1G4
Isotype: IgG1
Species Reactivity: Human. Others not known.
Epitope: aa 1-29
Immunogen: A synthetic peptide aa 1-29 (MAN-SASPEQNQNHCSAINNSIPLMQGNLPY) from the N-terminal of human GnRH receptor
Molecular Weight: 54-60 kDa
Positive Control: Pituitary or normal ovary
Cellular Localization: Membrane and cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Immunofluorescence
- Flow Cytometry
Note:
USA: RUO; Int'l: RUO
Specifications
GnRH Receptor / LH-RH Receptor Ab-3 | |
Monoclonal | |
200μg/mL | |
Purified Ab with BSA and Azide | |
Murine | |
IgG1 | |
RUO | |
Primary | |
Monoclonal |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
GNRH03; same as F1G4 | |
Unconjugated | |
Concentrated | |
A synthetic peptide aa 1-29 (MAN-SASPEQNQNHCSAINNSIPLMQGNLPY) from the N-terminal of human GnRH receptor | |
500μL | |
G Protein-Coupled Receptors | |
Primary | |
Human |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.