Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Goat IgG, AP, Isotype Control, Novus Biologicals™

Goat Isotype Control

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP196984

Catalog No. NBP196984


Add to cart

Description

Description

IgG antibody specifically detects IgG in Goat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, SDS-Page.
Specifications

Specifications

IgG
AP
0.05 M Tris Chloride, 0.15M Sodium Chloride, 0.001M Magnesium Chloride, 0.0001M Zinc Chloride, 50% (v/v) Glycerol; pH 8.0, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.1% Sodium Azide
immunoglobulin heavy constant gamma 1 (G1m marker)
Goat
Ion exchange chromatography
RUO
Isotype Control
3500
Goat
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), SDS-Page, Western Blot
Western Blot 1:500 - 1:2500, ELISA 1:2000 - 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, SDS-Page 1:100 - 1:2000
Conjugated
IGHG1
IgG
1 mg
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Primary
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Goat IgG, anti-Goat Serum and anti-Alkaline Phosphatase (calf intestine).
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only