Learn More
Seracare Life Sciences IncSupplier Diversity Partner ANTI-GOAT IGG (H+L) AB HRP  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 52200362
Affinity purified polyclonal antibody to Goat IgG, both heavy and light chains (whole IgG) made in Rabbit and labeled with horseradish peroxidase. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-672-24