Learn More
anti-CD28, Clone: 37.51, Novus Biologicals™
Golden Syrian Hamster Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP130418
Catalog No. NBP130418
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
CD28 Monoclonal antibody specifically detects CD28 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, Functional, CyTOF-ready.
Specifications
Specifications
CD28
37.51
Unconjugated
Phosphate buffered saline (PBS), approx. pH 7.4; 0.2 um filter sterilized. with No Preservative
CD28 antigen, CD28 antigen (Tp44), CD28 molecule, MGC138290, T-cell-specific surface glycoprotein CD28, Tp44
Golden Syrian Hamster
IgG
Protein G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
The Syrian hamster monoclonal antibody 37.51 reacts with extracellular domain of mouse CD28 costimulatory receptor, a 45 kDa homodimeric protein expressed by thymocytes, mature T cells and NK cells.
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen)
1.0 mg/mL
Flow Cytometry 6 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunoprecipitation 1-2 ug 100-500 ug of protein in 1 ml lysate, Immunohistochemistry-Frozen 1:200, Functional, CyTOF-ready
Purified
CD28
Mouse T cell lymphoma EL-4 cells
45 kDa
0.1 mg
Adaptive Immunity, Apoptosis, Immunology
Primary
940
Mouse