Learn More
anti-CD3 epsilon, Cy5, Clone: 500-A2, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP126684
Catalog No. NBP126684
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
CD3 epsilon
500-A2
Cyanine5
PBS with Sodium Azide
CD3e antigen, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, FLJ18683, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE
Golden Syrian Hamster
IgG Kappa
0.1 mg
Adaptive Immunity, Apoptosis, Diabetes Research, Immunology, Innate Immunity, Signal Transduction
Primary
916
Mouse
Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Frozen), In vitro assay
0.1 mg/mL
Flow Cytometry <= 1 ug/10^6 cells, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, In vitro assay
Purified
CD3E
Immune complexes isolated from C6VL-BS thymoma cells
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Mouse CD3 epsilon, Mr 25 kDa