Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-KLRG1, APC, Clone: 2F1, Novus Biologicals™

Golden Syrian Hamster Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP128115APC

Catalog No. NBP128115J


Add to cart

Description

Description

KLRG1 Monoclonal antibody specifically detects KLRG1 in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications

Specifications

KLRG1
2F1
Flow Cytometry
Purified
KLRG1
IL-2-activated NK cells (A-LAK cells) from B6 (H-2b) mice
Protein A or G purified
RUO
Store at 4C in the dark.
Monoclonal
KLRG1 (Mr 30-38 kDa)
Flow Cytometry
APC
PBS with 0.05% Sodium Azide
2F1, CLEC15AMAFA-LIKE, C-type lectin domain family 15 member A, C-type lectin domain family 15, member A, ITIM-containing receptor MAFA-L, killer cell lectin-like receptor subfamily G member 1, killer cell lectin-like receptor subfamily G, member 1, MAFAL, MAFA-L, MAFA-like receptor, MAFAMAFA-2F1, Mast cell function-associated antigen, mast cell function-associated antigen (ITIM-containing), MGC13600
Golden Syrian Hamster
IgG2
0.25 ml
Innate Immunity, Signal Transduction
Primary
10219
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only