Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Golgi protein 58k Goat anti-Human, Mouse, Porcine, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA131351

Catalog No. PIPA131351


Add to cart

Description

Description

PA1-31351 detects 58K Golgi protein from human samples. PA1-31351 has been successfully used in Western blot applications. The PA1-31351 immunogen is: Synthetic peptide: SQLVECVPNFSEGKNQ, corresponding to N terminal amino acids 2-17 of Human 58K Golgi protein. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

The protein encoded by this gene is a bifunctional enzyme that channels 1-carbon units from formiminoglutamate, a metabolite of the histidine degradation pathway, to the folate pool. Mutations in this gene are associated with glutamate formiminotransferase deficiency. Alternatively spliced transcript variants have been found for this gene.
Specifications

Specifications

Golgi protein 58k
Polyclonal
Unconjugated
FTCD
Liquid
FTCD
synthetic peptide SQLVECVPNFSEGKNQ, corresponding to N terminal amino acids 2-17 of Human 58K Golgi protein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Porcine
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
O95954, Q91XD4, P53603
LCHC1
Goat
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
10841, 14317, 397517
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit