Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Golgin-84 Mouse, Unlabeled, Clone: 26, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611383

 View more versions of this product

Catalog No. BDB611383


Add to cart

Description

Description

The Golgi apparatus is a complex and dynamic organelle that functions in protein sorting and modification. Numerous structural and regulatory proteins are involved in the budding, docking, and fusion of Golgi-directed vesicles. Golgin-84 is an integral membrane protein associated with the Golgi. Sequence analysis of the C-terminal region of Golgin-84 demonstrates a 14 residue region extending into the lumen of the Golgi, a membrane insertion sequence, and a Golgi retention signal. The large N-terminal cytoplasmic region contains a coiled-coil domain that is required for dimerization and two leucine zipper domains. Golgin-84 is ubiquitously expressed, but is abundant in testis. Golgin-84 has sequence homology to coiled-coil containing myosin family members and to several other Golgi proteins, including trans-Golgi p230 and gigantin. Trans-Golgi p230 has been implicated in the biogenesis of specific Golgi vesicles, while gigantin is implicated in the tethering of non-clathrin coated vesicles and in the anchoring of adjacent Golgi cisternae. Thus, Golgin-84 may have similar roles in vesicle biogenesis, docking, and fusion at the Golgi interface.

Immunofluorescence, Western Blotting

Specifications

Specifications

Golgin-84
26
Unconjugated
Affinity Purified
Human Golgin-84 aa. 510-713
150μg
Cell Biology
Primary
Canine, Human
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.