Learn More
Golgin-97 Mouse anti-Human, Clone: CDF4, Invitrogen™ Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen A21270
Catalog No. A21270
Add to cart
Description
Description
Anti-human golgin-97 antibodies were originally isolated from the serum of a patient with the autoimmune disease known as Sjögren ft.s syndrome. These antibodies recognize a 97 kDa protein called golgin-97, a member of the granin family of proteins and a peripheral membrane protein localized on the cytoplasmic face of the Golgi apparatus. Because the antibody recognizes a protein unique to the Golgi apparatus of most vertebrate species, it is useful for immunodetection and identification of the Golgi apparatus in cells. To prepare a stock solution, reconstitute the antibody in 0.5-1 mL of phosphate-buffered saline, pH 7.4, containing 1% BSA. Store the solution for up to two weeks at 4°C with the addition of 2 mM sodium azide. For longer storage, divide the solution into single-use aliquots and freeze at -20°C.
The Golgi apparatus, which participates in glycosylation and transport of proteins and lipids in the secretory pathway, consists of a series of stacked cisternae (flattened membrane sacs). Interactions between the Golgi and microtubules are thought to be important for the reorganization of the Golgi after it fragments during mitosis. The golgins are a family of proteins, of which the protein encoded by this gene is a member, that are localized to the Golgi. This encoded protein is associated with Sjogren ft.s syndrome.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Golgin-97
Monoclonal
Unconjugated
GOLGA1
Lyophilized
GOLGA1
Human golgin-97.
Purified
RUO
Antibody
Monoclonal
Human
Immunocytochemistry, Immunofluorescence
CDF4
Antibody with no preservative
Q92805
golgin-97, Golgin subfamily A member 1, gap junction protein, alpha 4, 37kD, golgi autoantigen, golgin subfamily a, 1
Mouse
IgG1
100 μg
-20°C
Primary
2800