Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GOLPH3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA514347

Catalog No. PIPA514347

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with mouse and rat based on sequence homology.

The Golgi complex plays a key role in the sorting and modification of proteins exported from the endoplasmic reticulum. This protein is a peripheral membrane protein of the Golgi stack and may have a regulatory role in Golgi trafficking.
Specifications

Specifications

GOLPH3
Polyclonal
PBS with 0.09% sodium azide
Q9H4A6
Coat protein GPP34; Golgi phosphoprotein 3; GOLPH3; GPP34
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
64083
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
GOLPH3
Liquid
GOLPH3
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human GOLPH3
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit