Learn More
anti-gp130, Alexa Fluor 700, Clone: 28126, R&D Systems™
Manufacturer:  R&D Systems FAB228N100
 View more versions of this product
Catalog No. FAB228N100
Add to cart
Specifications
Specifications
gp130/CD130
28126
Flow Cytometry 5 uL/10^6 cells
Purified
IL6ST
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human gp130 extracellular domain
Protein A or G purified from ascites
RUO
Primary
3572
Human
Flow Cytometry
Alexa Fluor 700
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
CD130, CD130 antigen, CDw130, DKFZp564F053, gp130, gp130 of the rheumatoid arthritis antigenic peptide-bearing soluble form, IL-6 receptor subunit beta, IL-6R subunit beta, IL-6RB, IL-6R-beta, IL6ST, interleukin 6 signal transducer (gp130, oncostatin M receptor), interleukin receptor beta chain, interleukin-6 receptor subunit beta, Interleukin-6 signal transducer, Membrane glycoprotein 130, membrane glycoprotein gp130, Oncostatin-M receptor subunit alpha
Mouse
IgG1
100 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Monoclonal
Detects human gp130 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) IL-2 R, rhIL-4 R, or rhIL-6 R is observed.