Learn More
glycoprotein Ib (platelet), alpha polypeptide, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human GP1BA protein.
Supplier: Abnova Corporation H00002811D01P
Description
Glycoprotein Ib (GP Ib) is a platelet surface membrane glycoprotein composed of a heterodimer, an alpha chain and a beta chain, that are linked by disulfide bonds. The Gp Ib functions as a receptor for von Willebrand factor (VWF). The complete receptor complex includes noncovalent association of the alpha and beta subunits with platelet glycoprotein IX and platelet glycoprotein V. The binding of the GP Ib-IX-V complex to VWF facilitates initial platelet adhesion to vascular subendothelium after vascular injury, and also initiates signaling events within the platelet that lead to enhanced platelet activation, thrombosis, and hemostasis. This gene encodes the alpha subunit. Several polymorphisms and mutations have been described in this gene, some of which are the cause of Bernard-Soulier syndromes and platelet-type von Willebrand disease. [provided by RefSeq
Sequence: MPLLLLLLLLPSPLHPHPICEVSKVASHLEVNCDKRNLTALPPDLPKDTTILHLSENLLYTFSLATLMPYTRLTQLNLDRCELTKLQVDGTLPVLGTLDLSHNQLQSLPLLGQTLPALTVLDVSFNRLTSLPLGALRGLGELQELYLKGNELKTLPPGLLTPTPKLEKLSLANNNLTELPAGLLNGLENLDTLLLQENSLYTIPKGFFGSHLLPFAFLHGNPWLCNCEILYFRRWLQDNAENVYVWKQGVDVKAMTSNVASVQCDNSDKFPVYKYPGKGCPTLGDEGDTDLYDYYPEEDTEGDKVRATRTVVKFPTKAHTTPWGLFYSWSTASLDSQMPSSLHPTQESTKEQTTFPPRWTPNFTLHMESITFSKTPKSTTEPTPSPTTSEPVPEPAPNMTTLEPTPSPTTPEPTSEPAPSPTTPEPTPIPTIATSPTILVSATSLITPKSTFLTTTKPVSLLESTKKTIPELDQPPKLRGVLQGHLESSRNDPFLHPDFCCLLPLGFYVLGLFWLLFASVVLILLLSWVGHVKPQALDSGQGAALTTATQTTHLELQRGRQVTVPRAWLLFLRGSLPTFRSSLFLWVRPNGRLGPLVAGRRPSALSQGRGQDLLSTVSIRYSGHSLSpecifications
glycoprotein Ib (platelet), alpha polypeptide | |
Polyclonal | |
Rabbit polyclonal antibody raised against a full-length human GP1BA protein. | |
GP1BA | |
BSS/CD42B/CD42b-alpha/GP1B/MGC34595 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
Antibody |
In situ PLA, Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC027955 | |
GP1BA | |
GP1BA (AAH27955.1, 1 a.a. ∼ 626 a.a) full-length human protein. | |
100 μg | |
Cell Adhesion | |
2811 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.